Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-methylbenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Numéro de catalogue. AAH62717MD
Modifier l'affichage
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAH62717MD 250 mg
AAH6271703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAH62717MD Fournisseur Thermo Scientific Chemicals Code fournisseur H62717MD

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1001200-60-7
Molecular Formula C13H18BFO2
Molecular Weight (g/mol) 236.093
MDL Number MFCD18729904
InChI Key UNTVLIKQVDPHFS-UHFFFAOYSA-N
Synonym 2-4-fluoro-3-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-3-methylphenylboronic acid pinacol ester, 4-fluoro-3-methylphenylboronic acid, pinacol ester, amtb633
PubChem CID 56776823
IUPAC Name 2-(4-fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C

Spécifications

Quantity 250 mg
Formula Weight 236.09
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Fluoro-3-methylbenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.