Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-methylbenzenesulfonyl chloride, 97%, Thermo Scientific™

Catalog No. AAH6411003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6411003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6411003 Supplier Thermo Scientific Chemicals Supplier No. H6411003

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629672-19-1
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.63
MDL Number MFCD01365818
InChI Key ALEQALYEZFNWOP-UHFFFAOYSA-N
Synonym 4-fluoro-3-methyl-benzenesulfonyl chloride, 4-fluoro-3-methylbenzenesulfonylchloride, 4-fluoro-3-methylbenzene-1-sulfonyl chloride, 4-fluoro-3-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro-3-methyl, chloro 4-fluoro-3-methylphenyl sulfone, ablock ab-12-8863, 5-chlorosulphonyl-2-fluorotoluene, 4-fluro-3-methylphenyl-sulfonylchloride, 4-fluoro-3-methylphenyl-sulfonylchloride
PubChem CID 2782758
SMILES CC1=C(F)C=CC(=C1)S(Cl)(=O)=O

Specifications

Melting Point 35°C to 40°C
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3261
Beilstein 3254603
Sensitivity Moisture sensitive
IUPAC Name 4-fluoro-3-methylbenzenesulfonyl chloride
Formula Weight 208.63
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Fluoro-3-methylbenzenesulfonyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.