Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-nitrobenzaldehyde, 97%, Thermo Scientific™

Catalog No. SB02056DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB02056DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB02056DA Supplier Thermo Scientific Chemicals Supplier No. SB02056DA
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 42564-51-2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H4FNO3
Linear Formula FC6H3(NO2)CHO
Quantity 1 g
MDL Number MFCD01861388
Synonym 3-nitro-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzaldehyde, pubchem22572, acmc-1ao2q, 3-nitro-4 fluorobenzaldehyde, ksc497m9r, 2-fluoro-5-formylnitrobenzene, #, 4-fluoranyl-3-nitro-benzaldehyde
InChI Key ILKWFRCNNILIJW-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C=O)[N+](=O)[O-])F
IUPAC Name 4-fluoro-3-nitrobenzaldehyde
Molecular Weight (g/mol) 169.111
PubChem CID 598129
Formula Weight 169.11
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-nitrobenzaldehyde
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.