Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH6272006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6272006 5 g
AAH6272003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6272006 Supplier Thermo Scientific Chemicals Supplier No. H6272006

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Fluoro-3-nitrobenzeneboronic acid is generally used as a intermediate in pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1218791-09-3
Molecular Formula C12H15BFNO4
MDL Number MFCD12546515
Synonym 2-3-fluoro-4-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-fluoro-4-nitrophenylboronic acid pinacol ester, 3-fluoro-4-nitrophenylboronic acid,pinacol ester, 3-fluoro-4-nitrophenylboronic acid, pinacol ester, 2-3-fluoro-4-nitrophenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane
PubChem CID 46739531

Specifications

Melting Point 64°C to 65°C
Color Yellow
Quantity 5 g
Beilstein 20289173
Solubility Information Slightly soluble in water.
InChI Key QBMHYMHOXGOPAF-UHFFFAOYSA-N
SMILES CC1(C)OB(OC1(C)C)C1=CC(=C(F)C=C1)[N+]([O-])=O
IUPAC Name 2-(4-fluoro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol) 267.06
Formula Weight 267.06
Percent Purity 96%
Physical Form Crystalline Solid
Chemical Name or Material 4-Fluoro-3-nitrobenzeneboronic acid pinacol ester
Show More Show Less
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.