Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-nitrobenzotrifluoride, 97%

Catalog No. AAA1967614
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1967614 25 g
AAA1967622 100 g
AAA1967636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1967614 Supplier Thermo Scientific Chemicals Supplier No. A1967614
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 367-86-2 | C7H3F4NO2 | 209.1 g/mol

4-Fluoro-3-nitrobenzotrifluoride is used in the pre-column derivatization technique for HPLC with UV/VIS spectrophotometric detection of polyamines. It may be used as derivatization reagent for the HPLC determination of polyamines. It was also used in the synthesis of 2-nitro-4-trifluoromethylphenylhydrazine, derivatization reagent for 1,2-diol compounds and oxo-steroids.

  • [3-(2-nitro-4-trifluoromethylphenyl)aminophenyl]dihydroxyborane, derivatization reagent for 1,2-diol compounds and oxo-steroids3
  • 1-(3-chlorophenyl)-5-trifluoromethyl-3-hydrobenzimidazol-2-one

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-3-nitrobenzotrifluoride is used in the pre-column derivatization technique for HPLC with UV/VIS spectrophotometric detection of polyamines. It may be used as derivatization reagent for the HPLC determination of polyamines. It was also used in the synthesis of 2-nitro-4-trifluoromethylphenylhydrazine, derivatization reagent for 1,2-diol compounds and oxo-steroids. • [3-(2-nitro-4-trifluoromethylphenyl)aminophenyl]dihydroxyborane, derivatization reagent for 1,2-diol compounds and oxo-steroids3 • 1-(3-chlorophenyl)-5-trifluoromethyl-3-hydrobenzimidazol-2-one

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-86-2
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD00007059
InChI Key HLDFCCHSOZWKAA-UHFFFAOYSA-N
Synonym 4-fluoro-3-nitrobenzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl benzene, benzene, 1-fluoro-2-nitro-4-trifluoromethyl, 4-fluoro-3-nitrotrifluoromethylbenzene, 3-nitro-4-fluorobenzotrifluoride, 3-nitro-alpha,alpha,alpha,4-tetrafluorotoluene, 2-fluoro-5-trifluoromethyl nitrobenzene, 3-nitro-4-fluoro benzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl-benzene
PubChem CID 67778
IUPAC Name 1-fluoro-2-nitro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])F

Specifications

Density 1.494
Boiling Point 91°C to 92°C (15 mmHg)
Flash Point 90°C (194°F)
Refractive Index 1.462
Quantity 25 g
UN Number UN2810
Beilstein 1880508
Solubility Information Insoluble in water.
Formula Weight 209.1
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-nitrobenzotrifluoride
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H311-H227-H302-H332-H315-H319
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 206-702-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.