Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-phenoxybenzaldehyde, 97%, Thermo Scientific™

Catalog No. AAH6120206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6120206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6120206 Supplier Thermo Scientific Chemicals Supplier No. H6120206

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68359-57-9
Molecular Formula C13H9FO2
Molecular Weight (g/mol) 216.21
MDL Number MFCD01318148
InChI Key JDICMOLUAHZVDS-UHFFFAOYSA-N
Synonym 4-fluoro-3-phenoxy-benzaldehyde, benzaldehyde, 4-fluoro-3-phenoxy, m-phenoxy-p-fluorobenzaldehyde, 2-fluoro-5-formyldiphenyl ether, 4-fluoro-3-phenoxy benzaldehyde, pubchem14130, 3-phenoxy-4fluorobenzaldehyde, ksc497q9b, 3-phenoxy-4-fluorobenzaldehyde, 3-phenoxy-4-fluoro-benzaldehyde
PubChem CID 110068
IUPAC Name 4-fluoro-3-phenoxybenzaldehyde
SMILES FC1=CC=C(C=O)C=C1OC1=CC=CC=C1

Specifications

Color Colorless
Density 1.209 g/mL
Flash Point >110°C (230°F)
Refractive Index 1.583
Quantity 5 g
UN Number UN3082
Formula Weight 216.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Fluoro-3-phenoxybenzaldehyde
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 269-856-2
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.