Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Fluoro-4-(8-fluoro1,3,4,5tetrahydro-2H-pyrido[4,3b]indol2yl)butyrophenone HCl 99%, Thermo Scientific™

Catalog No. AC182690250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC182690250 25 g
AC182690050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC182690250 Supplier Thermo Scientific Chemicals Supplier No. 182690250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31540-62-2
Molecular Formula C21H21ClF2N2O
Molecular Weight (g/mol) 390.859
InChI Key LGNFRLBPCVLGOB-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl-4-8-fluoro-2,3,4,5-tetrahydro-1h-pyrido 4,3-b indol-2-ium-2-yl butan-1-one chloride, 4-8-fluoro 1,2,3,4-tetrahydropyridino 4,3-b indol-2-yl-1-4-fluorophenyl bu tan-1-one, chloride, regid_for_cid_24213095, 1-4-fluorophenyl-4-8-fluoro-2,3,4,5-tetrahydro-1h-pyrido, 1-4-fluorophenyl-4-8-fluoro-1,3,4,5-tetrahydro-2h-pyrido 4,3-b indol-2-yl-1-butanone hydrochloride, 4-8-fluoro-3,4-dihydro-1h-pyrido 4,3-b indol-2 5h-yl-1-4-fluorophenyl butan-1-one hydrochloride, 4-8-fluoro-1h,3h,4h,5h-pyrido 4,3-b indol-2-yl-1-4-fluorophenyl butan-1-one hydrochloride
PubChem CID 24213095
IUPAC Name 1-(4-fluorophenyl)-4-(8-fluoro-1,3,4,5-tetrahydropyrido[4,3-b]indol-2-yl)butan-1-one;hydrochloride
SMILES C1CN(CC2=C1NC3=C2C=C(C=C3)F)CCCC(=O)C4=CC=C(C=C4)F.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Quantity 25 g
Formula Weight 390.85
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 4'-Fluoro-4-(8-fluoro-1,3,4,5-tetrahydro-2H-pyrido[4,3-b]indol-2-yl)butyrophenoneHCl, 99%
EINECSNumber 250-688-3
RTECSNumber EU2625000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.