Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-7-nitrobenzofurazan

Catalog No. AAJ61336MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61336MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61336MC Supplier Thermo Scientific Chemicals Supplier No. J61336MC
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 29270-56-2 | C6H2FN3O3 | 183.098 g/mol

4-Chloro-7-nitrobenzofurazan (NBD-chloride) was used in the following studies, Synthesis of fluorescent phospholipid-derivative, NBD-didecanoylphosphatidylethanolamine, Synthesis of functionalized hydroxynaphthofurazan, Spectrophotometric and spectrofluorometric determination of clemastine hydrogen fumarate, loratadine, losartan potassium and ramipril in pharmaceutical formulations. Synthesis of 7-nitrobenzofurazan (NBD)-labeled maleimide, via Diels-Alder reaction. NBD-Cl is widely used to label peptides, proteins, drugs and other biomolecules. It is also a popular derivatizing reagent for HPLC analysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-7-nitrobenzofurazan (NBD-chloride) was used in the following studies, Synthesis of fluorescent phospholipid-derivative, NBD-didecanoylphosphatidylethanolamine, Synthesis of functionalized hydroxynaphthofurazan, Spectrophoto
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29270-56-2
Molecular Formula C6H2FN3O3
Molecular Weight (g/mol) 183.098
MDL Number MFCD00010196
InChI Key PGZIDERTDJHJFY-UHFFFAOYSA-N
Synonym 4-fluoro-7-nitrobenzofurazan, nbd-f, 4-fluoro-7-nitrobenzo c 1,2,5 oxadiazole, 7-fluoro-4-nitrobenzofurazan, 7-fluoro-4-nitrobenzo-2-oxa-1,3-diazole, 4-fluoro-7-nitrobenz-2-oxa-1,3-diazole, 2,1,3-benzoxadiazole, 4-fluoro-7-nitro, nbd-fluoride, nbd-f cpd, 4-fluoro-7-nitrobenzo c 1,2,5-oxadiazole
PubChem CID 122123
IUPAC Name 4-fluoro-7-nitro-2,1,3-benzoxadiazole
SMILES C1=C(C2=NON=C2C(=C1)F)[N+](=O)[O-]

Specifications

Melting Point 53°C to 57°C
Color Yellow
Flash Point 110°C (230°F)
Quantity 100 mg
Beilstein 1077571
Solubility Information It is soluble in DMSO,DMF,CH3OH,CH3CN and CHCl3.
Formula Weight 183.1
Physical Form Powder
Chemical Name or Material 4-Fluoro-7-nitrobenzofurazan
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.