Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Fluoro-alpha-methylbenzyl alcohol

Catalog No. 1327055G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-270-55G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-270-55G Supplier Sigma Aldrich Supplier No. 1327055G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Fluoro-alpha-methylbenzyl alcohol

Specifications

CAS 403-41-8
Density 1.109 g/mL (at 25°C (literature))
Boiling Point 90°C to 92°C (7 mmHg)
Molecular Formula C8H9FO
Refractive Index n20/D 1.501 (literature)
Linear Formula FC6H4CH(CH3)OH
Quantity 250 mg
MDL Number MFCD00004515
Molecular Weight (g/mol) 140.15
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.