Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-alpha-methylbenzyl Alcohol 97.0+%, TCI America™
SDP

Catalog No. F04325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04325G 5 g
F043225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04325G Supplier TCI America Supplier No. F04325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 403-41-8
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.16
MDL Number MFCD00004515,MFCD03092997,MFCD03092998
InChI Key PSDSORRYQPTKSV-UHFFFAOYNA-N
Synonym 1-4-fluorophenyl ethanol, 4-fluoro-alpha-methylbenzyl alcohol, 1-4-fluorophenyl ethan-1-ol, 1-p-fluorophenyl ethanol, p-fluorophenylmethylcarbinol, 4-fluorophenylmethylcarbinol, 1-4-fluorophenyl-1-ethanol, alpha-methyl-4-fluorobenzyl alcohol, 1-4-fluorophenyl-ethanol, 4-fluorophenyl methyl carbinol
PubChem CID 73946
IUPAC Name 1-(4-fluorophenyl)ethan-1-ol
SMILES CC(O)C1=CC=C(F)C=C1

Specifications

Color Colorless
Boiling Point 123°C
Quantity 5 g
Formula Weight 140.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-alpha-methylbenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.