Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-D-2-phenylglycine 98.0+%, TCI America™
SDP

Catalog No. F08635G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08635G 5 g
F08631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08635G Supplier TCI America Supplier No. F08635G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 93939-74-3
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.155
MDL Number MFCD00042727
InChI Key JKFYKCYQEWQPTM-SSDOTTSWSA-N
Synonym r-4-fluorophenylglycine, r-2-amino-2-4-fluorophenyl acetic acid, d--4-fluorophenylglycine, 4-fluoro-d-phenylglycine, 4-fluoro-d-alpha-phenylglycine, d-4-fluorophenylglycine, r---4-fluorophenyl glycine, 4-fluoro-d-2-phenylglycine, 4-fluorophenylglycine, 4-fluoro-d-a-phenylglycine
PubChem CID 124515
IUPAC Name (2R)-2-amino-2-(4-fluorophenyl)acetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)N)F

Specifications

Color White
Quantity 5 g
Formula Weight 169.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-D-2-phenylglycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.