Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-D-phenylalanine 98.0+%, TCI America™
SDP

Catalog No. F09011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09011G 1 g
F09015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09011G Supplier TCI America Supplier No. F09011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18125-46-7
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.182
MDL Number MFCD00063063
InChI Key XWHHYOYVRVGJJY-MRVPVSSYSA-N
Synonym 4-fluoro-d-phenylalanine, d-4-fluorophenylalanine, h-d-phe 4-f-oh, r-2-amino-3-4-fluorophenyl propanoic acid, p-fluoro-d-phenylalanine, d-phenylalanine, 4-fluoro, 2r-2-amino-3-4-fluorophenyl propanoic acid, 4-fluorophenylalanine, d, unii-963g7o1pve, r-4-fluorophenylalanine
PubChem CID 716314
IUPAC Name (2R)-2-amino-3-(4-fluorophenyl)propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)F

Specifications

Melting Point 247°C
Color White
Quantity 1 g
Formula Weight 183.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-D-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.