Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-L-phenylglycine, 98%, Thermo Scientific™

Catalog No. AAH6149514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6149514 25 g
AAH6149522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6149514 Supplier Thermo Scientific Chemicals Supplier No. H6149514

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Fluoro-L-phenylglycine is an optically active amino acid derivative that is used as an intermediate for making drugs.

Solubility
Soluble in 1M HCl.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19883-57-9
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.16
MDL Number MFCD00213799
InChI Key JKFYKCYQEWQPTM-ZETCQYMHSA-N
Synonym s-4-fluorophenylglycine, s-2-amino-2-4-fluorophenyl acetic acid, 4-fluoro-l-alpha-phenylglycine, l-4-fluorophenylglycine, 4-fluoro-l-phenylglycine, l +-4-fluorophenylglycine, 2s-2-amino-2-4-fluorophenyl acetic acid, s-+-4-fluorophenyl glycine, p-fluorophenyl glycine, 4-fluorophenylglycine
PubChem CID 853015
SMILES [NH3+][C@H](C([O-])=O)C1=CC=C(F)C=C1

Specifications

Melting Point >300°C
Color White
Beilstein 5850631
Solubility Information Soluble in 1M HCl.
IUPAC Name (2S)-2-azaniumyl-2-(4-fluorophenyl)acetate
Quantity 25 g
Formula Weight 169.15
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Fluoro-L-phenylglycine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.