Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-N-Fmoc-L-phenylalanine, 95%

Catalog No. AAH5197203
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5197203 1 g
AAH51972MD 250 mg
AAH5197206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5197203 Supplier Thermo Scientific Chemicals Supplier No. H5197203
Only null left
Add to Cart

CAS: 169243-86-1 | C24H20FNO4 | 405.43 g/mol

4-Fluoro-N-Fmoc-L-phenylalanine derivative was introduced in collaboration with Yu and coworkers in reflection to a methodology reported in C-H Activation. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-N-Fmoc-L-phenylalanine derivative was introduced in collaboration with Yu and coworkers in reflection to a methodology reported in C-H Activation. It is also used as pharmaceutical intermediate.

Solubility
Soluble in Dimethylformamide(DMF).

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169243-86-1
Molecular Formula C24H20FNO4
Molecular Weight (g/mol) 405.43
MDL Number MFCD00191197
InChI Key IXUMACXMEZBPJG-QFIPXVFZSA-N
Synonym fmoc-4-fluoro-l-phenylalanine, fmoc-l-4-fluorophenylalanine, fmoc-phe 4-f-oh, fmoc-l-4-fluorophe, fmoc-4-fluoro-l-phe, fmoc-4-fluoro-l-phe-oh, fmoc-l-phe 4-f-oh, fmoc-p-fluoro-phe-oh, l-4-fluoro-phenyl-n-fmoc-alanine, s-n-fmoc-4-fluorophenylalanine
PubChem CID 2734466
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-fluorophenyl)propanoic acid
SMILES OC(=O)[C@H](CC1=CC=C(F)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 1 g
Beilstein 8287377
Solubility Information Soluble in Dimethylformamide(DMF).
Formula Weight 405.42
Percent Purity 95%
Chemical Name or Material 4-Fluoro-N-Fmoc-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.