Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Fluoroaniline

Catalog No. F3800500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F3800500G 500 g
F380025G 25 g
F3800100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F3800500G Supplier Sigma Aldrich Supplier No. F3800500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Fluoroaniline

Specifications

CAS 371-40-4
Density 1.173 g/mL (at 25°C (literature))
Boiling Point 187°C (767 mmHg)
Molecular Formula C6H6FN
Refractive Index n20/D 1.539 (literature)
Linear Formula FC6H4NH2
MDL Number MFCD00007829
Quantity 500 g
Synonym 1-Amino-4-fluorobenzene
Molecular Weight (g/mol) 111.12
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.