Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzamide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
824-75-9
C7H6FNO
139.13
MFCD00007992
VNDHYTGVCGVETQ-UHFFFAOYSA-N
p-fluorobenzamide, benzamide, 4-fluoro, benzamide, p-fluoro, p-fluorobenzoic acid amide, 4-fluoro-benzamide, unii-yb79f3xb1g, yb79f3xb1g, 4fluorobenzamide, 4-fluorobenzoamide, pubchem3407
71572
4-fluorobenzamide
C1=CC(=CC=C1C(=O)N)F
824-75-9
C7H6FNO
139.13
MFCD00007992
VNDHYTGVCGVETQ-UHFFFAOYSA-N
p-fluorobenzamide, benzamide, 4-fluoro, benzamide, p-fluoro, p-fluorobenzoic acid amide, 4-fluoro-benzamide, unii-yb79f3xb1g, yb79f3xb1g, 4fluorobenzamide, 4-fluorobenzoamide, pubchem3407
71572
4-fluorobenzamide
C1=CC(=CC=C1C(=O)N)F
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Melting Point 154.0°C to 158.0°C
Color White to Beige
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula FC6H4CONH2
Beilstein 09, I, 137
Formula Weight 139.13
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4-Fluorobenzamide, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-535-9

RTECSNumber : CV4956666