Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzil 98.0+%, TCI America™
SDP

Catalog No. F0989200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0989200MG 200 mg
F09891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0989200MG Supplier TCI America Supplier No. F0989200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3834-66-0
Molecular Formula C14H9FO2
Molecular Weight (g/mol) 228.222
MDL Number MFCD00223483
InChI Key JKQPFVOJZNRINB-UHFFFAOYSA-N
Synonym 4-Fluorobibenzoyl, 4-Fluorodibenzoyl
PubChem CID 3625310
IUPAC Name 1-(4-fluorophenyl)-2-phenylethane-1,2-dione
SMILES C1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)F

Specifications

Melting Point 64°C
Quantity 200 mg
Formula Weight 228.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorobenzil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.