Learn More
Medchemexpress LLC 4-Fluorobenzoic acid | 456-22-4 | MFCD00002530 | 98.8% | 140.11 | 250 G
SDP
Catalog No. 5000453408
Encompass_Preferred
4-Fluorobenzoic acid is a versatile drug intermediate. It serves as a key building block for synthesizing compounds with important biological activities.
  • Synthesizes hydrazone derivatives for antituberculosis applications.
  • Synthesizes Schiff bases with DPPH radical scavenging activity.
Catalog No. 50-004-53408 Supplier Medchemexpress LLC Supplier No. 250GHYW013677
Add to Cart
Add to Cart
4-Fluorobenzoic acid is a versatile drug intermediate. It serves as a key building block for synthesizing compounds with important biological activities.
  • Synthesizes hydrazone derivatives for antituberculosis applications.
  • Synthesizes Schiff bases with DPPH radical scavenging activity.