Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzyl alcohol, 97%, Thermo Scientific™

Catalog No. AAA1233618
Change view
Click to view available options
Quantity:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1233618 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1233618 Supplier Thermo Scientific Chemicals Supplier No. A1233618

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Fluorobenzyl alcohol is used in the preparation of many compounds including amides. It is also used as an pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and acid anhydrides.
TRUSTED_SUSTAINABILITY

Specifications

CAS 459-56-3
Density 1.161
Boiling Point 206°C
Flash Point 90°C (194°F)
Odor Characteristic
Molecular Formula C7H7FO
Refractive Index 1.508
Quantity 50g
MDL Number MFCD00004651
Beilstein 1446930
Synonym 4-fluorobenzyl alcohol, 4-fluorophenyl methanol, benzenemethanol, 4-fluoro, p-fluorobenzyl alcohol, 4-fluorobenzylalcohol, 4-fluorobenzylic alcohol, ccris 5115, 4-fluorobenzoic alcohol, 4-fluoro benzyl alcohol, 4-fluorophenyl methan-1-ol
Solubility Information Slightly soluble in water.
InChI Key GEZMEIHVFSWOCA-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CO)F
IUPAC Name (4-fluorophenyl)methanol
Molecular Weight (g/mol) 126.13
PubChem CID 68022
Formula Weight 126.13
Percent Purity 97%
Chemical Name or Material 4-Fluorobenzyl alcohol
Show More Show Less
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 207-292-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.