Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzyl triphenyl phosphoniumchloride, 98%, Thermo Scientific™

Catalog No. AC330790250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC330790250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC330790250 Supplier Thermo Scientific Chemicals Supplier No. 330790250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Specifications

CAS 3462-95-1
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C25H21ClFP
Quantity 25g
MDL Number MFCD00031637
Synonym 4-fluorobenzyl triphenylphosphonium chloride, phosphonium, 4-fluorophenyl methyl triphenyl-, chloride, 4-fluorophenyl methyl triphenylphosphanium chloride, acmc-20a9k1, ksc495g9t, guljcsointwknf-uhfffaoysa-m, 4-fluorobenzyl triphenylphosphine chloride, p-fluorobenzyltriphenylphosphonium chloride, 4-fluorobenzyltriphenyl phosphonium chloride
InChI Key CBHDAHHYMRXLIP-UHFFFAOYSA-M
SMILES C1=CC=C(C=C1)[P+](CC2=CC=C(C=C2)F)(C3=CC=CC=C3)C4=CC=CC=C4.[Cl-]
IUPAC Name (4-fluorophenyl)methyl-triphenylphosphanium;chloride
Molecular Weight (g/mol) 406.865
PubChem CID 2733554
Formula Weight 406.87
Percent Purity ≥97.5%
Chemical Name or Material 4-Fluorobenzyl triphenyl phosphoniumchloride, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.