Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroacetone, 96%, stabilized

Catalog No. AC173470010
Change view
Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173470010 1 L Glass bottle
AC173470050 5 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC173470010 Supplier Thermo Scientific Chemicals Supplier No. 173470010

CAS: 78-95-5 | C3H5ClO | 92.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-95-5
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
MDL Number MFCD00000936
InChI Key BULLHNJGPPOUOX-UHFFFAOYSA-N
Synonym chloroacetone, chloropropanone, acetonyl chloride, monochloroacetone, tonite, 2-propanone, 1-chloro, chloro-2-propanone, 1-chloroacetone, chloromethyl methyl ketone, 1-chloro-2-propanone
PubChem CID 6571
ChEBI CHEBI:47220
IUPAC Name 1-chloropropan-2-one
SMILES CC(=O)CCl

Specifications

Melting Point -44.5°C
Color Brown-Yellow to Yellow
Density 1.1600g/mL
Boiling Point 119.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4310 to 1.4330
Linear Formula ClCH2COCH3
Quantity 1 L
Beilstein 01, 653
Specific Gravity 1.16
Merck Index 15, 2114
Solubility Information Solubility in water: 124g/L (20°C). Other solubilities: soluble in alcohol, ether and chloroform
Stabilizer 0.1% epoxidized soybean oil
Viscosity 1.11 mPa.s (25°C)
Formula Weight 92.52
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Chloroacetone, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Fatal if inhaled.
Causes ski
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
EINECSNumber 201-161-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.