Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzylmagnesium chloride, 0.25M solution in THF, AcroSeal™

Catalog No. AC426790500
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC426790500 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC426790500 Supplier Thermo Scientific Chemicals Supplier No. 426790500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1643-73-8 | C7H6ClFMg | 168.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1643-73-8
Molecular Formula C7H6ClFMg
Molecular Weight (g/mol) 168.88
MDL Number MFCD01319898
InChI Key PQDGQUPDDGUKLP-UHFFFAOYSA-M
Synonym 4-fluorobenzylmagnesium chloride, chloro 4-fluorophenyl methyl magnesium, 4-fluorobenzylmagnesiumchloride, 4-fluorobenzyl magnesium chloride, pqdgqupddguklp-uhfffaoysa-m, 4-fluoro benzylmagnesium chloride, 4-fluorobenzylmagnesium chloride, 0.25 m in thf, 4-fluorobenzylmagnesium chloride 0.25 m in tetrahydrofuran, 4-fluorobenzylmagnesium chloride, 0.25m w/w in thf
PubChem CID 2778244
IUPAC Name magnesium;1-fluoro-4-methanidylbenzene;chloride
SMILES FC1=CC=C(C[Mg]Cl)C=C1

Specifications

Color Brown or Orange to Yellow
CAS Min % 2.67
CAS Max % 6.24
Linear Formula FC6H4CH2MgCl
Concentration 0.15 to 0.35M (exact strength on the certificate of analysis)
Density 0.9480g/mL
Formula Weight 168.88
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.948
Quantity 50 mL
Physical Form Solution
Chemical Name or Material 4-Fluorobenzylmagnesium chloride, 0.25M solution in THF
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May cause respiratory irritation. Causes skin irritation. Causes serious eye damage. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. If skin irritation occurs: Get medical advice/attention. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.