Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorodiphenylmethane 95.0+%, TCI America™
SDP

Catalog No. F05685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05685G Supplier TCI America Supplier No. F05685G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 587-79-1
Molecular Formula C13H11F
Molecular Weight (g/mol) 186.23
MDL Number MFCD00464329
InChI Key ADCBAIUWZPOIMC-UHFFFAOYSA-N
PubChem CID 604241
IUPAC Name 1-benzyl-4-fluorobenzene
SMILES FC1=CC=C(CC2=CC=CC=C2)C=C1

Specifications

Color Yellow
Boiling Point 132°C
Quantity 5 g
Formula Weight 186.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluorodiphenylmethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.