Learn More
Cayman Chemical 4-Fluoroethcathinone (hydrochloride), Purity: ≥98%, CAS: 2705473-25-0, 5 mg
SDP
Catalog No. 502951191
Encompass_Preferred
A structural analog of 4-FMC, with an ethyl group replacing the amino terminal methyl group; intended to be used for research and forensic applications
Catalog No. 50-295-1191 Supplier Cayman Chemical Supplier No. 112315 MG
A structural analog of 4-FMC, with an ethyl group replacing the amino terminal methyl group; intended to be used for research and forensic applications