Learn More
Cayman Chemical 4-Fluoromethcathinone metabolite (hydrochloride) ((±)-Ephedrine stereochemistry), Purity: ≥98%, CAS: 7799-72-6, 1 mg
SDP
Catalog No. 502972694
Encompass_Preferred
A metabolite of 4-FMC that features conversion of the β-keto group to β-hydroxy and is an enantiomeric mixture of the R,S and S,R orientations at carbons one and two, as in ephedrine; intended for forensic and research purposes
Catalog No. 50-297-2694 Supplier Cayman Chemical Supplier No. 147571 MG
A metabolite of 4-FMC that features conversion of the β-keto group to β-hydroxy and is an enantiomeric mixture of the R,S and S,R orientations at carbons one and two, as in ephedrine; intended for forensic and research purposes