Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenyl Acetate 98.0+%, TCI America™
SDP

Catalog No. A14295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14295G Supplier TCI America Supplier No. A14295G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 405-51-6
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00041361
InChI Key ZNOREXRHKZXVPC-UHFFFAOYSA-N
Synonym 1-acetoxy-4-fluorobenzene, 4-fluorophenyl acetate, p-fluorophenyl acetate, acetic acid 4-fluorophenyl ester, phenol,4-fluoro-,acetate, 4-fluorophenylacetate, pubchem3061, acmc-1acsq, ksc493c7p, phenol, 4-fluoro-, 1-acetate
PubChem CID 136253
IUPAC Name (4-fluorophenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)F

Specifications

Color Yellow
Boiling Point 86°C
Quantity 5 g
Formula Weight 154.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluorophenyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.