Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluorophenylhydrazine hydrochloride, 98%

Click to view available options
Quantity:
5g
25g
100g
823-85-8
C6H8ClFN2
162.592
MFCD00012942
FEKUXLUOKFSMRO-UHFFFAOYSA-N
4-fluorophenylhydrazine hydrochloride,4-fluorophenyl hydrazine hydrochloride,4-fluorophenylhydrazine hcl,1-4-fluorophenyl hydrazine hydrochloride,4-fluorophenyl hydrazine hcl,p-fluorophenylhydrazine hydrochloride,4-fluorphenyl hydrazine hydrochloride,4-fluorophenyl hydrazine monohydrochloride,p-fluorophenylhydrazine hcl,hydrazine, 4-fluorophenyl-, monohydrochloride
69981
(4-fluorophenyl)hydrazine;hydrochloride
C1=CC(=CC=C1NN)F.Cl
823-85-8
C6H8ClFN2
162.592
MFCD00012942
FEKUXLUOKFSMRO-UHFFFAOYSA-N
4-fluorophenylhydrazine hydrochloride,4-fluorophenyl hydrazine hydrochloride,4-fluorophenylhydrazine hcl,1-4-fluorophenyl hydrazine hydrochloride,4-fluorophenyl hydrazine hcl,p-fluorophenylhydrazine hydrochloride,4-fluorphenyl hydrazine hydrochloride,4-fluorophenyl hydrazine monohydrochloride,p-fluorophenylhydrazine hcl,hydrazine, 4-fluorophenyl-, monohydrochloride
69981
(4-fluorophenyl)hydrazine;hydrochloride
C1=CC(=CC=C1NN)F.Cl
Applications
4-Fluorophenylhydrazine hydrochloride is used as a chemical intermediate in the pharmaceutica1ls particularly of tryptamine drugs used in the treatment of migraine and cluster headaches. It is used to prepare agrochemical, and other chemical. It is used to detect sugars and aldehydes in analytical chemistry.

Solubility
Soluble in water.

Notes
Store in cool dry place in tightly closed container. Store away from oxidizing agent.
Quantity 5g
Melting Point ~265°C (decomposition)
Beilstein 3627721
Solubility Information Soluble in water.
Formula Weight 162.6
Percent Purity 98%
Chemical Name or Material 4-Fluorophenylhydrazine hydrochloride

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P362-P501c

H302+H312+H332-H315-H317-H319-H335-H351

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 212-521-2

TSCA : No

Recommended Storage : Ambient temperatures