Learn More
eMolecules​ 352-13-6 | 4-Fluorophenylmagnesium bromide, 1.0 M in THF | Synthonix - Stock | MFCD00009667 | 199.305 | C6H4BrFMg | 0.000 | [Br-].Fc1ccc([Mg+])cc1 | 100ml | 437237224
Catalog No. 502697620 Shop All eMolecules Inc. Products
Encompass_Preferred
4-Fluorophenylmagnesium bromide, 1.0 M in THF | Synthonix - Stock | 352-13-6 | MFCD00009667 | 199.305 | C6H4BrFMg | 0.000 | [Br-].Fc1ccc([Mg+])cc1 | 100ml | 437237224
Catalog No. 50-269-7620 Supplier eMolecules​ Supplier No. 437237224
4-Fluorophenylmagnesium bromide, 1.0 M in THF | Synthonix - Stock | 352-13-6 | MFCD00009667 | 199.305 | C6H4BrFMg | 0.000 | [Br-].Fc1ccc([Mg+])cc1 | 100ml | 437237224