Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenylmagnesium Bromide (ca. 19% in Tetrahydrofuran, ca. 1.0mol/L), TCI America™
SDP

Catalog No. F0673250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0673250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F0673250G Supplier TCI America Supplier No. F0673250G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 352-13-6
Molecular Formula C6H4BrFMg
Molecular Weight (g/mol) 199.31
MDL Number MFCD00009667
InChI Key QYBFFRXNNFXREA-UHFFFAOYSA-M
Synonym 4-fluorophenylmagnesium bromide, magnesium, bromo 4-fluorophenyl, bromo 4-fluorophenyl magnesium, p-fluorophenylmagnesium bromide, 4-fluorophenyl magnesium bromide, 4-f-c6h4-mgbr, 4-fluorophenylmagnesiumbromide, 4-fluorphenylmagnesium bromide, 4-flourophenylmagnesium bromide, 4-fluorophenyl magnesiumbromide
PubChem CID 2734897
IUPAC Name bromo(4-fluorophenyl)magnesium
SMILES FC1=CC=C([Mg]Br)C=C1

Specifications

Quantity 250 g
UN Number 2924
Formula Weight 199.31
Chemical Name or Material 4-Fluorophenylmagnesium Bromide (ca. 19% in Tetrahydrofuran, ca. 1.0mol/L)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.