Learn More
Medchemexpress LLC 4-Fluororesorcinol | 103068-41-3 | 128.10 | 1 ML
SDP
Catalog No. 5000448610
Encompass_Preferred
4-Fluororesorcinol is a precursor utilized in the creation of fluorescent dyes or indicators, including compounds like fluorescein, coumarin, and various resorcinol analogues.
  • Precursor for fluorescent dyes and indicators.
  • Used in the preparation of fluorescein.
  • Used in the preparation of coumarin.
  • Used in the preparation of resorcinol analogues.
Catalog No. 50-004-48610 Supplier Medchemexpress LLC Supplier No. HYW00311210MM/1ML
4-Fluororesorcinol is a precursor utilized in the creation of fluorescent dyes or indicators, including compounds like fluorescein, coumarin, and various resorcinol analogues.
  • Precursor for fluorescent dyes and indicators.
  • Used in the preparation of fluorescein.
  • Used in the preparation of coumarin.
  • Used in the preparation of resorcinol analogues.