Learn More
Medchemexpress LLC 4-Fluororesorcinol | 103068-41-3 | 128.10 | 500 MG
SDP
Catalog No. 5000445602
Encompass_Preferred
4-Fluororesorcinol is a precursor used in the preparation of fluorescent dyes or indicators. It is suitable for research applications, contributing to the synthesis of compounds such as fluorescein, coumarin, and resorcinol analogues.
  • Precursor for fluorescent dyes and indicators
  • Suitable for research use only
  • Used in the preparation of fluorescein, coumarin, and resorcinol analogues
Catalog No. 50-004-45602 Supplier Medchemexpress LLC Supplier No. HYW003112500MG
4-Fluororesorcinol is a precursor used in the preparation of fluorescent dyes or indicators. It is suitable for research applications, contributing to the synthesis of compounds such as fluorescein, coumarin, and resorcinol analogues.
  • Precursor for fluorescent dyes and indicators
  • Suitable for research use only
  • Used in the preparation of fluorescein, coumarin, and resorcinol analogues