Learn More
Medchemexpress LLC 4-Fluororesorcinol | 103068-41-3 | MFCD03789074 | 128.10 | 100 MG
SDP
Catalog No. 5000449140
Encompass_Preferred
4-Fluororesorcinol is a chemical compound used as a precursor in the preparation of various fluorescent dyes and indicators. It plays a role in synthesizing compounds such as fluorescein, coumarin, and resorcinol analogues, which are widely utilized in scientific research and analytical applications.
  • Precursor for fluorescent dyes
  • Used in synthesis of fluorescein
  • Applied in coumarin production
  • Key in creating resorcinol analogues
  • Valuable for scientific research
  • Essential for analytical applications
Catalog No. 50-004-49140 Supplier Medchemexpress LLC Supplier No. HYW003112100MG
4-Fluororesorcinol is a chemical compound used as a precursor in the preparation of various fluorescent dyes and indicators. It plays a role in synthesizing compounds such as fluorescein, coumarin, and resorcinol analogues, which are widely utilized in scientific research and analytical applications.
  • Precursor for fluorescent dyes
  • Used in synthesis of fluorescein
  • Applied in coumarin production
  • Key in creating resorcinol analogues
  • Valuable for scientific research
  • Essential for analytical applications