Learn More
Medchemexpress LLC 4'-Fluorouridine | 1613589-24-4 | MFCD33551704 | 99.9% | 262.19 | C9H11FN2O6 | 5 MG
SDP
Catalog No. 5000222295
Encompass_Preferred
EIDD-2749 (4'-Fluorouridine) is a ribonucleoside analog and orally active RNA-dependent RNA polymerase (RdRp) inhibitor used for antiviral research. It has demonstrated activity against respiratory syncytial virus, SARS-CoV-2, hepatitis C virus, and lymphocytic choriomeningitis virus, and is supplied at research-grade purity for laboratory studies.
  • Orally active RdRp inhibitor.
  • Demonstrated antiviral activity against multiple RNA viruses.
  • High purity suitable for research applications.
  • Available in small milligram pack sizes for dose-ranging studies.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-22295 Supplier Medchemexpress LLC Supplier No. HY1462465MG
EIDD-2749 (4'-Fluorouridine) is a ribonucleoside analog and orally active RNA-dependent RNA polymerase (RdRp) inhibitor used for antiviral research. It has demonstrated activity against respiratory syncytial virus, SARS-CoV-2, hepatitis C virus, and lymphocytic choriomeningitis virus, and is supplied at research-grade purity for laboratory studies.
  • Orally active RdRp inhibitor.
  • Demonstrated antiviral activity against multiple RNA viruses.
  • High purity suitable for research applications.
  • Available in small milligram pack sizes for dose-ranging studies.
  • Stable when stored under recommended conditions.