Learn More
Medchemexpress LLC 4'-Fluorouridine triphosphate sodium | 97.0% | 568.08 | 100 UL
SDP
Catalog No. 5000363857
Encompass_Preferred
4'-Fluorouridine triphosphate sodium is a bioactive 5'-triphosphate form of 4'-Fluorouridine (EIDD-2749). It effectively binds to RSV RdRP and is intended for research use only.
  • Bioactive 5'-triphosphate form of 4'-Fluorouridine
  • Effectively binds to RSV RdRP
  • For research use only
Catalog No. 50-003-63857 Supplier Medchemexpress LLC Supplier No. HY146247A5.68MG
4'-Fluorouridine triphosphate sodium is a bioactive 5'-triphosphate form of 4'-Fluorouridine (EIDD-2749). It effectively binds to RSV RdRP and is intended for research use only.
  • Bioactive 5'-triphosphate form of 4'-Fluorouridine
  • Effectively binds to RSV RdRP
  • For research use only