Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical (4-Fmoc-amino-tetrahydrothiopyran-4-yl)-acetic acid | 10 g | CAS 946727-62-4 | MDL MFCD09264207

Catalog No. 502352182
Encompass

(4-Fmoc-amino-tetrahydrothiopyran-4-yl)-acetic acid is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 946727-62-4
- MDL: MFCD09264207
- InChIKey: MOZSRIGQQHPMTM-UHFFFAOYSA-N
- Molecular Weight: 397.489
- Molecular Formula: C22H23NO4S
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: 2-(4-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)tetrahydro-2H-thiopyran-4-yl)acetic acid
- SMILES: O=C(CC1(NC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)CCSCC1)O

Catalog No. 50-235-2182 Supplier STA Pharmaceutical Supplier No. STAAA1011010GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(4-Fmoc-amino-tetrahydrothiopyran-4-yl)-acetic acid is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 946727-62-4
- MDL: MFCD09264207
- InChIKey: MOZSRIGQQHPMTM-UHFFFAOYSA-N
- Molecular Weight: 397.489
- Molecular Formula: C22H23NO4S
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: 2-(4-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)tetrahydro-2H-thiopyran-4-yl)acetic acid
- SMILES: O=C(CC1(NC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)CCSCC1)O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.