Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA PHARMACEUTICAL US LLC 4-Fmoc-piperazine-2-carboxylic acid | 5 g | CAS 219312-90-0 | MDL MFCD01076312

Catalog No. 502345511
Encompass

4-Fmoc-piperazine-2-carboxylic acid is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 219312-90-0
- MDL: MFCD01076312
- InChIKey: MVXOLCRCKMWZCB-UHFFFAOYSA-N
- Molecular Weight: 352.39
- Molecular Formula: C20H20N2O4
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29335995
- Country Of Origin: China
- IUPAC: 4-(((9H-fluoren-9-yl)methoxy)carbonyl)piperazine-2-carboxylic acid
- SMILES: O=C(N1CCNC(C(O)=O)C1)OCC2C3=CC=CC=C3C4=CC=CC=C42

Catalog No. 50-234-5511 Supplier STA PHARMACEUTICAL US LLC Supplier No. STAAA115365GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4-Fmoc-piperazine-2-carboxylic acid is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 219312-90-0
- MDL: MFCD01076312
- InChIKey: MVXOLCRCKMWZCB-UHFFFAOYSA-N
- Molecular Weight: 352.39
- Molecular Formula: C20H20N2O4
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29335995
- Country Of Origin: China
- IUPAC: 4-(((9H-fluoren-9-yl)methoxy)carbonyl)piperazine-2-carboxylic acid
- SMILES: O=C(N1CCNC(C(O)=O)C1)OCC2C3=CC=CC=C3C4=CC=CC=C42

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.