Learn More
Electron Microscopy Sciences 4" Forceps with Straight Tips, Stainless Steel
SDP
Catalog No. 5029022
Encompass_Preferred
Made from stainless steel, medium points, serrated tips and alignment pin, curved or straight tips. Only curved type A tips comes with locking device.
Tips: Straight Tips
Catalog No. 50-290-22 Supplier Electron Microscopy Sciences Supplier No. 729914S
Made from stainless steel, medium points, serrated tips and alignment pin, curved or straight tips. Only curved type A tips comes with locking device.
Tips: Straight Tips