Learn More
Medchemexpress LLC 4-formylbenzoic acid | 619-66-9 | MFCD00006951 | 150.13 g·mol⁻¹ | C8H6O3 | 1 ML
SDP
Catalog No. 5000301780
Encompass_Preferred
4-formylbenzoic acid is an aromatic carboxylic aldehyde used as an intermediate in organic synthesis and as a reagent in materials and coordination chemistry. It participates in aldehyde condensation reactions and can be used to prepare functionalized aromatics and metal-organic coordination polymers. Supplied for research use as a solution to simplify handling in small-scale experiments.
  • Used as an intermediate in organic synthesis.
  • Participates in aldehyde condensation and derivatization reactions.
  • Can form coordination polymers with metal salts such as barium(II).
  • Useful for synthesis of acid-functionalized mesoporous silica catalysts.
  • Available as a convenient solution in DMSO for small-scale experiments.
  • Molecular formula C8H6O3; molecular weight 150.13 g·mol⁻¹.
Catalog No. 50-003-01780 Supplier Medchemexpress LLC Supplier No. HYW01744610MM/1ML
4-formylbenzoic acid is an aromatic carboxylic aldehyde used as an intermediate in organic synthesis and as a reagent in materials and coordination chemistry. It participates in aldehyde condensation reactions and can be used to prepare functionalized aromatics and metal-organic coordination polymers. Supplied for research use as a solution to simplify handling in small-scale experiments.
  • Used as an intermediate in organic synthesis.
  • Participates in aldehyde condensation and derivatization reactions.
  • Can form coordination polymers with metal salts such as barium(II).
  • Useful for synthesis of acid-functionalized mesoporous silica catalysts.
  • Available as a convenient solution in DMSO for small-scale experiments.
  • Molecular formula C8H6O3; molecular weight 150.13 g·mol⁻¹.