Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 4-Formylbenzoic acid, 619-66-9, MFCD00006951, 25g

Catalog No. 50681076
Encompass_Preferred

Molecular Formula C8H6O3, Purity 95+%, Molecular Weight 150.14, Melting Point ca 245°

Catalog No. 50-681-076 Supplier Matrix Scientific Supplier No. 07509625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C8H6O3, Purity 95+%, Molecular Weight 150.14, Melting Point ca 245°

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.