Learn More
Justrite™ 4 Gallon Sure-Grip™ EX Countertop Corrosives/Acid Steel Safety Cabinet With 1 Self-Closing Door
GREENER_CHOICE
Catalog No. 19822393 Shop All Justrite Safety Group Products
Change view
Click to view available options
Material:
Steel (CR)
Catalog No. Material
19-822-393 Steel (CR)
1 options
Catalog No. 19-822-393 Supplier Justrite™ Supplier No. 890422
  • $1,880.00
    $1,563.65 / Each of 1
    Save $316.35
    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 4 Gallon Sure-Grip™ EX Countertop Corrosives/Acid Steel Safety Cabinet With 1 Self-Closing Door is specially designed for safely storing acids and corrosives in laboratories.
Countertop and Compac Cabinets let you customize your work areas and make the most of available space. Whether adding to existing storage space or making chemicals readily available at workstation locations, these smaller-sized cabinets are the solution when space is at a premium.
Specifications
Product Type Safety Cabinet
Dimensions (D x W x H) 17 x 17 x 22 in.
Description 4 gallon safety cabinet with self-closing door; 18 Ga CR Steel
Material Steel (CR)