Learn More
Med Vet International 4" German Feline Declaw Forceps, Stainless Steel, J1026, Each
Catalog No. 5000585777
Encompass_Preferred
These forceps are very useful in feline declaws especially when doing the procedure with a laser. The wide end grabs the claw firmly so a precise incision can be made.
Catalog No. 50-005-85777 Supplier Med Vet International Supplier No. JOR1026
These forceps are very useful in feline declaws especially when doing the procedure with a laser. The wide end grabs the claw firmly so a precise incision can be made.