Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Heptanoylbiphenyl, TCI America™
SDP

Catalog No. H070825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H070825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H070825G Supplier TCI America Supplier No. H070825G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59662-27-0
Molecular Formula C19H22O
Molecular Weight (g/mol) 266.384
MDL Number MFCD00143347
InChI Key YCPKHSFFVZYFMP-UHFFFAOYSA-N
Synonym 4-Enanthoylbiphenyl, 4-Heptanoyldiphenyl
PubChem CID 108796
IUPAC Name 1-(4-phenylphenyl)heptan-1-one
SMILES CCCCCCC(=O)C1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 25 g
Formula Weight 266.38
Physical Form Crystalline Powder
Chemical Name or Material 4-Heptanoylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.