Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Heptylbenzoic Acid 95.0+%, TCI America™
SDP

Catalog No. H077410G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H077410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H077410G Supplier TCI America Supplier No. H077410G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38350-87-7
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.312
MDL Number MFCD00009722
InChI Key VSUKEWPHURLYTK-UHFFFAOYSA-N
Synonym benzoic acid, 4-heptyl, 4-n-heptylbenzoic acid, 4-heptylbenzoicacid, 4-heptylbenzoate, p-heptylbenzoic acid, acmc-209izm, 4-n-heptyl-benzoic acid, 4-heptylbenzoic acid
PubChem CID 170036
IUPAC Name 4-heptylbenzoic acid
SMILES CCCCCCCC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 102°C
Color White
Quantity 10 g
Formula Weight 220.31
Physical Form Crystalline Powder
Chemical Name or Material 4-Heptylbenzoic Acid
EINECSNumber (3)-1338
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.