Learn More
Medchemexpress LLC 4-Heptyloxyphenol | 13037-86-0 | 99.7% | 208.30 | 500 MG
SDP
Catalog No. 5000401290
Encompass_Preferred
4-Heptyloxyphenol, also known as p-(Heptyloxy)phenol, is a chemical compound that exhibits antibacterial activity against specific bacteria.
  • Antibacterial activity
  • Active against P. gingivalis
  • Active against S. artemidis
  • Active against Str. sobrinus
  • MIC values: 0.10, 0.21, 0.14 mM
Catalog No. 50-004-01290 Supplier Medchemexpress LLC Supplier No. HY30263500MG
4-Heptyloxyphenol, also known as p-(Heptyloxy)phenol, is a chemical compound that exhibits antibacterial activity against specific bacteria.
  • Antibacterial activity
  • Active against P. gingivalis
  • Active against S. artemidis
  • Active against Str. sobrinus
  • MIC values: 0.10, 0.21, 0.14 mM