Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hexadecyloxybenzoic acid, 98%, Thermo Scientific™

Catalog No. AC208301000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC208301000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC208301000 Supplier Thermo Scientific Chemicals Supplier No. 208301000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 15872-48-7
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C23H38O3
Linear Formula CH3(CH2)15OC6H4CO2H
Quantity 100g
MDL Number MFCD00002544
Beilstein 10, III, 286
Synonym 4-hexadecyloxybenzoic acid, 4-hexadecyloxy benzoic acid, p-hexadecyloxybenzoic acid, 4-n-hexadecyloxybenzoic acid, benzoic acid, 4-hexadecyloxy, p-hexadecyloxy benzoic acid, 4-hexadecyloxy-benzoic acid, benzoic acid,4-hexadecyloxy
InChI Key GAQCVRTXIAGNEM-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCOC1=CC=C(C=C1)C(=O)O
IUPAC Name 4-hexadecoxybenzoic acid
Molecular Weight (g/mol) 362.554
PubChem CID 259635
Formula Weight 362.54
Percent Purity ≥97.5%
Chemical Name or Material 4-Hexadecyloxybenzoic acid, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.