Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hexen-1-ol, predominantly trans, 97%, Thermo Scientific Chemicals

Catalog No. p-7036201
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5445906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5445906 Supplier Thermo Scientific Chemicals Supplier No. H5445906
Only null left
Add to Cart

4-Hexen-1-ol on reaction with NaIO4/NaHSO3 reagent yields 2-(1-iodoethyl)tetrahydrofuran. has been employed in the preparation of hex-4-enals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hexen-1-ol on reaction with NaIO4/NaHSO3 reagent yields 2-(1-iodoethyl)tetrahydrofuran. has been employed in the preparation of hex-4-enals.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-92-7
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00009713
InChI Key VTIODUHBZHNXFP-NSCUHMNNSA-N
Synonym 4-hexen-1-ol, e-4-hexen-1-ol, e-hex-4-en-1-ol, trans-4-hexen-1-ol, 4-hexen-1-ol, 4e, 4-hexen-1-ol, e, trans-4-hexenol, unii-zhb2532c2h, 4-hexen-1-ol, predominantly trans
PubChem CID 641248
IUPAC Name (E)-hex-4-en-1-ol
SMILES C\C=C\CCCO

Specifications

Density 0.851
Boiling Point 159°C to 160°C
Flash Point 61°C (142°F)
Refractive Index 1.439
Linear Formula CH3CH=CH(CH2)3OH
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 100.16
Percent Purity 97%
Chemical Name or Material 4-Hexen-1-ol, predominantly trans
Hazard Category H227-H319
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
EINECSNumber 213-188-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.