Learn More
Medchemexpress LLC 4-[[(hexylamino)carbonyl]amino]-N-[4-[2-[[(2S)-2-hydroxy-3-(4-hydroxyphenoxy)propyl]amino]ethyl]-phe | 159182-43-1 | 98.9% | 584.73 g/mol | C30H40N4O6S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1933410MG
Encompass_Preferred
This compound is a potent, selective β3-adrenergic receptor agonist (IC50 35 nM) reported to enhance homology-directed repair (HDR)-mediated genome editing. It is supplied as a high-purity solid with characterized molecular and storage properties suitable for pharmacology and genome-editing research.
  • Potent β3-adrenergic receptor agonist with IC50 of 35 nM.
  • Enhances homology-directed repair (HDR) for genome-editing applications.
  • High purity suitable for research use.
  • Solid, white to off-white appearance for easy handling.
  • Stable storage profile for powder and for solutions.
Catalog No. 50-002-98390