Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hexyloxyphenol 98.0+%, TCI America™
SDP

Catalog No. H04395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04395G 5 g
H043925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04395G Supplier TCI America Supplier No. H04395G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18979-55-0
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.27
MDL Number MFCD00002337
InChI Key XIIIHRLCKLSYNH-UHFFFAOYSA-N
Synonym 4-hexyloxyphenol, 4-hexyloxy phenol, phenol, 4-hexyloxy, 4-n-hexyloxyphenol, p-hexyloxyphenol, unii-35ims9l1fe, 35ims9l1fe, pubchem13208, acmc-1c6hg, hydroquinone monohexyl ether
PubChem CID 29354
ChEBI CHEBI:34407
IUPAC Name 4-(hexyloxy)phenol
SMILES CCCCCCOC1=CC=C(O)C=C1

Specifications

Melting Point 46°C
Color White-Yellow
Boiling Point 112°C
Quantity 5 g
Formula Weight 194.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hexyloxyphenol
EINECSNumber (3)-3539
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.