Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Hexylresorcinol

Catalog No. 20946525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-946-525G 25 g
20-946-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-946-525G Supplier Sigma Aldrich Supplier No. 20946525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Hexylresorcinol

Specifications

CAS 136-77-6
Melting Point 65°C to 67°C (lit.)
Boiling Point 333°C to 335°C (lit.)
Molecular Formula C12H18O2
Linear Formula CH3(CH2)5C6H3-1,3-(OH)2
MDL Number MFCD00002284
Quantity 25 g
Synonym Antascarin; Ascarinol; 4-Hexyl-1,3-dihydroxybenzene
Molecular Weight (g/mol) 194.27
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.