Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hexylthiophene-2-carboxaldehyde 97.0+%, TCI America™
SDP

Catalog No. H14991G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H14991G Supplier TCI America Supplier No. H14991G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 222554-30-5
Molecular Formula C11H16OS
Molecular Weight (g/mol) 196.31
MDL Number MFCD18414651
InChI Key XZSDWKHOEIKBDB-UHFFFAOYSA-N
Synonym 2-Formyl-4-hexylthiophene
PubChem CID 58420936
IUPAC Name 4-hexylthiophene-2-carbaldehyde
SMILES CCCCCCC1=CSC(C=O)=C1

Specifications

Quantity 1 g
Formula Weight 196.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Hexylthiophene-2-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.